Learn More
Meso Scale Discovery S-PLEX Human IL-3 KitQP
SDP
Catalog No. 5000079584
Encompass
The S-PLEX Human IL-3 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human interleukin 3 (IL-3) in serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection technology The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label assay diluent calibrator and wash buffer The assay has a sensitivity of 0 08 pg/mL and an upper limit of quantification of 2500 pg/mL for human IL-3 This multiplex kit provides high sensitivity requires low sample volume and allows for rapid results
Catalog No. 50-000-79584 Supplier Meso Scale Discovery Supplier No. K151ADQS21
The S-PLEX Human IL-3 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human interleukin 3 (IL-3) in serum plasma and cell culture supernatants using electrochemiluminescence (ECL) detection technology The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label assay diluent calibrator and wash buffer The assay has a sensitivity of 0 08 pg/mL and an upper limit of quantification of 2500 pg/mL for human IL-3 This multiplex kit provides high sensitivity requires low sample volume and allows for rapid results