Learn More
Meso Scale Discovery S-PLEX Human IL-3 KitQP
SDP
Catalog No. 5000080365
Encompass
The Meso Scale Discovery S-PLEX Human Kinexus Profiling Assay Kit is a multiplex immunoassay that simultaneously measures the levels of 151 human protein biomarkers It utilizes electrochemiluminescent detection technology and provides a rapid sensitive and reproducible method for profiling a broad range of human proteins in serum plasma and cell culture supernatants The assay requires a small sample volume and provides quantitative data on important biomarkers related to cancer immunology neurobiology and cardiovascular disease research The kit includes a 10-plate kit with reagents calibrators controls plates and accessories needed to run the assay
Catalog No. 50-000-80365 Supplier Meso Scale Discovery Supplier No. K151ADQS22
The Meso Scale Discovery S-PLEX Human Kinexus Profiling Assay Kit is a multiplex immunoassay that simultaneously measures the levels of 151 human protein biomarkers It utilizes electrochemiluminescent detection technology and provides a rapid sensitive and reproducible method for profiling a broad range of human proteins in serum plasma and cell culture supernatants The assay requires a small sample volume and provides quantitative data on important biomarkers related to cancer immunology neurobiology and cardiovascular disease research The kit includes a 10-plate kit with reagents calibrators controls plates and accessories needed to run the assay