Learn More
Meso Scale Discovery S-PLEX Human IL-5 KitQP
SDP
Catalog No. 5000082478
Encompass
The Meso Scale Discovery S-PLEX Human IL-5 Kit is a multiplex immunoassay that enables simultaneous quantitative measurement of IL-5 biomarker in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology on patterned arrays for high sensitivity and a wide dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents optimized for human samples The assay has high sensitivity with an LLOQ of 0 08 pg/mL and requires only 25 L of sample
Catalog No. 50-000-82478 Supplier Meso Scale Discovery Supplier No. K151J3S21
The Meso Scale Discovery S-PLEX Human IL-5 Kit is a multiplex immunoassay that enables simultaneous quantitative measurement of IL-5 biomarker in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology on patterned arrays for high sensitivity and a wide dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluents optimized for human samples The assay has high sensitivity with an LLOQ of 0 08 pg/mL and requires only 25 L of sample