Learn More
Meso Scale Discovery S-PLEX Human IL-6 Kit
SDP
Catalog No. 5000079623
Encompass
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatant samples The assay has high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete The kit has a detection range of 0 06-1500 pg/mL and includes reagents for 1 x 96-well plate
Catalog No. 50-000-79623 Supplier Meso Scale Discovery Supplier No. K151B3S1
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatant samples The assay has high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent wash buffer read buffer and plate seals The assay takes approximately 3 5 hours to complete The kit has a detection range of 0 06-1500 pg/mL and includes reagents for 1 x 96-well plate