Learn More
Meso Scale Discovery S-PLEX Human IL-6 KitQP
SDP
Catalog No. 5000080163
Encompass
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that enables the simultaneous detection of the human cytokine interleukin 6 (IL-6) using electrochemiluminescence (ECL) technology on the MSD platform This multiplex assay kit includes a plate coated with capture antibodies detection antibodies conjugated to an ECL label calibrators and diluent The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for IL-6 It is suitable for measuring IL-6 levels in human serum plasma and cell culture supernatants The kit provides components for 1 plate with 96 wells that can assay 40 samples in duplicate
Catalog No. 50-000-80163 Supplier Meso Scale Discovery Supplier No. K151B3S21
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that enables the simultaneous detection of the human cytokine interleukin 6 (IL-6) using electrochemiluminescence (ECL) technology on the MSD platform This multiplex assay kit includes a plate coated with capture antibodies detection antibodies conjugated to an ECL label calibrators and diluent The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for IL-6 It is suitable for measuring IL-6 levels in human serum plasma and cell culture supernatants The kit provides components for 1 plate with 96 wells that can assay 40 samples in duplicate