Learn More
Meso Scale Discovery S-PLEX Human IL-6 KitQP
SDP
Catalog No. 5000082040
Encompass
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that enables the simultaneous detection of the human cytokine interleukin 6 (IL-6) using electrochemiluminescent technology This multiplex assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG labels calibrators and an MSD Read Buffer for performing the assay The kit has a detection range of 0 05-1500 pg/mL and requires 50 L of sample per well It is compatible with serum plasma cell culture supernatants and other biological sample types The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-82040 Supplier Meso Scale Discovery Supplier No. K151B3S22
The Meso Scale Discovery S-PLEX Human IL-6 Kit is an immunoassay that enables the simultaneous detection of the human cytokine interleukin 6 (IL-6) using electrochemiluminescent technology This multiplex assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG labels calibrators and an MSD Read Buffer for performing the assay The kit has a detection range of 0 05-1500 pg/mL and requires 50 L of sample per well It is compatible with serum plasma cell culture supernatants and other biological sample types The assay takes approximately 3 5 hours to complete