Learn More
Meso Scale Discovery S-PLEX Human IL-9 Kit
SDP
Catalog No. 5000078366
Encompass
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay for detecting and quantifying human interleukin-9 (IL-9) It uses MSD s electrochemiluminescent technology and is compatible with the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate with IL-9 specific capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions for a standard curve It detects 0 06-2500 pg/mL with minimal cross-reactivity aiding cytokine profiling and immune response characterization in human serum plasma and cell culture supernatants
This item is not returnable. View return policy
Catalog No. 50-000-78366 Supplier Meso Scale Discovery Supplier No. K151R3S2
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay for detecting and quantifying human interleukin-9 (IL-9) It uses MSD s electrochemiluminescent technology and is compatible with the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate with IL-9 specific capture antibodies detection antibodies with an electrochemiluminescent label and calibrator solutions for a standard curve It detects 0 06-2500 pg/mL with minimal cross-reactivity aiding cytokine profiling and immune response characterization in human serum plasma and cell culture supernatants