Learn More
Meso Scale Discovery S-PLEX Human IL-9 Kit
SDP
Catalog No. 5000082194
Encompass
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay that allows simultaneous measurement of IL-9 biomarker levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation period The kit can test up to 80 samples in duplicate and has a shelf life of 9 months when stored at 2-8 C
Catalog No. 50-000-82194 Supplier Meso Scale Discovery Supplier No. K151R3S1
Add to Cart
Add to Cart
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay that allows simultaneous measurement of IL-9 biomarker levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation period The kit can test up to 80 samples in duplicate and has a shelf life of 9 months when stored at 2-8 C