Learn More
Meso Scale Discovery S-PLEX Human IL-9 Kit 25 PL
SDP
Catalog No. 5000078797
Encompass
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay that enables simultaneous quantitative measurement of IL-9 biomarker in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-78797 Supplier Meso Scale Discovery Supplier No. K151R3S4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay that enables simultaneous quantitative measurement of IL-9 biomarker in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation