Learn More
Meso Scale Discovery S-PLEX Human IL-9 KitQP
SDP
Catalog No. 5000078509
Encompass
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay that enables simultaneous quantitative measurement of IL-9 biomarker in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The assay has a detection range of 0 06-2500 pg/mL and requires 50 L of sample The S-PLEX platform combines electrochemiluminescence with patterned arrays providing high sensitivity up to 5 logs of dynamic range and low sample consumption This IL-9 assay kit allows for efficient and reproducible quantification of the IL-9 biomarker
Catalog No. 50-000-78509 Supplier Meso Scale Discovery Supplier No. K151R3S22
Add to Cart
Add to Cart
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay that enables simultaneous quantitative measurement of IL-9 biomarker in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The assay has a detection range of 0 06-2500 pg/mL and requires 50 L of sample The S-PLEX platform combines electrochemiluminescence with patterned arrays providing high sensitivity up to 5 logs of dynamic range and low sample consumption This IL-9 assay kit allows for efficient and reproducible quantification of the IL-9 biomarker