Learn More
Meso Scale Discovery S-PLEX Human IL-9 KitQP
SDP
Catalog No. 5000079880
Encompass
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay for detecting and quantifying human interleukin-9 (IL-9) It uses MSD s SECTOR Imager and electrochemiluminescent detection for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies for IL-9 detection antibodies with an MSD SULFO-TAG label calibrators and assay diluent It has a sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL enabling rapid reproducible IL-9 quantification from various biological samples
Catalog No. 50-000-79880 Supplier Meso Scale Discovery Supplier No. K151R3S21
The Meso Scale Discovery S-PLEX Human IL-9 Kit is a multiplex immunoassay for detecting and quantifying human interleukin-9 (IL-9) It uses MSD s SECTOR Imager and electrochemiluminescent detection for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies for IL-9 detection antibodies with an MSD SULFO-TAG label calibrators and assay diluent It has a sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL enabling rapid reproducible IL-9 quantification from various biological samples