Learn More
Meso Scale Discovery S-PLEX Human Neurofilament L Kit, 5 plates, 96-well, for Neurofilament L detection, LLOD 1,700 fg/mL, Dynamic Range 1,700 - 1,400,000 fg/mL
SDP
Catalog No. NC3304660
Encompass
S-PLEX Human Neurofilament L Kit, 5 plates, 96-well, for Neurofilament L detection, LLOD 1,700 fg/mL, Dynamic Range 1,700 - 1,400,000 fg/mL
Catalog No. NC3304660 Supplier Meso Scale Discovery Supplier No. K151AKGS2
Add to Cart
Add to Cart
Request a Quote
S-PLEX Human Neurofilament L Kit, 5 plates, 96-well, for Neurofilament L detection, LLOD 1,700 fg/mL, Dynamic Range 1,700 - 1,400,000 fg/mL