Learn More
Meso Scale Discovery S-PLEX Human Tau Kit, Kit Size 5 plate size, Analyte Tau (total), Lower Limit Of Detection 37 fg/mL, Dynamic Range 37 - 207,000 fg/mL, Species Human, Mouse, Rat, Non-human primate, Plate Type 96-well, Sample Types Serum, Plasma, Cell Culture Supernatant, CSF
SDP
Catalog No. NC2733615
Encompass
S-PLEX Human Tau Kit, Kit Size 5 plate size, Analyte Tau (total), Lower Limit Of Detection 37 fg/mL, Dynamic Range 37 - 207,000 fg/mL, Species Human, Mouse, Rat, Non-human primate, Plate Type 96-well, Sample Types Serum, Plasma, Cell Culture Supernatant, CSF
Catalog No. NC2733615 Supplier Meso Scale Discovery Supplier No. K151APSS2
Add to Cart
Add to Cart
Request a Quote
S-PLEX Human Tau Kit, Kit Size 5 plate size, Analyte Tau (total), Lower Limit Of Detection 37 fg/mL, Dynamic Range 37 - 207,000 fg/mL, Species Human, Mouse, Rat, Non-human primate, Plate Type 96-well, Sample Types Serum, Plasma, Cell Culture Supernatant, CSF