Learn More
Meso Scale Discovery S-PLEX Human Tau PT231 Kit, single-analyte assay, 96-well plate format, sensitivity 3,100 fg/mL, dynamic range 3,100 - 15,000,000 fg/mL, for human samples
SDP
Catalog No. NC3739857
Encompass
S-PLEX Human Tau PT231 Kit, single-analyte assay, 96-well plate format, sensitivity 3,100 fg/mL, dynamic range 3,100 - 15,000,000 fg/mL, for human samples
Catalog No. NC3739857 Supplier Meso Scale Discovery Supplier No. K151AGNS2
Add to Cart
Add to Cart
Request a Quote
S-PLEX Human Tau PT231 Kit, single-analyte assay, 96-well plate format, sensitivity 3,100 fg/mL, dynamic range 3,100 - 15,000,000 fg/mL, for human samples