Learn More
Meso Scale Discovery S-PLEX Human Tau pT231 KitQP
SDP
Catalog No. 5000080594
Encompass
The S-PLEX Human Total Tau Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of total tau in human serum plasma and CSF The kit contains a 96-well plate coated with a monoclonal antibody to human total tau a ruthenium-conjugated detection antibody calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a range of 50-3200 pg/mL and takes 3 5 hours to complete It uses MSD s proprietary S-PLEX technology is optimized for neurology biomarker analysis and is for research use only The kit offers high sensitivity and requires only 25 L sample volume
Catalog No. 50-000-80594 Supplier Meso Scale Discovery Supplier No. K151AGNS22
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human Total Tau Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay for the quantitative measurement of total tau in human serum plasma and CSF The kit contains a 96-well plate coated with a monoclonal antibody to human total tau a ruthenium-conjugated detection antibody calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a range of 50-3200 pg/mL and takes 3 5 hours to complete It uses MSD s proprietary S-PLEX technology is optimized for neurology biomarker analysis and is for research use only The kit offers high sensitivity and requires only 25 L sample volume