Learn More
Meso Scale Discovery S-PLEX Human TNF-α Kit, 5 plate size, sensitivity 6.8 fg/mL, dynamic range 6.8 - 75,000 fg/mL, sample types human serum, EDTA plasma, citrate plasma, heparin plasma, cell culture supernatants
SDP
Catalog No. NC3355616
Encompass
The MESO SCALE DISCOVERY S-PLEX Human TNF-α Kit is designed for the quantitative detection of human Tumor Necrosis Factor-alpha (TNF-α) with ultra sensitivity. Utilizing advanced electrochemiluminescence technology, this kit drastically improves sensitivity and dynamic range, aiming to facilitate biomarker research. Its straightforward protocol and compatibility with various sample types make it an essential tool in research labs.
  • Assay platform utilizes advanced technology
  • High sensitivity, achieving 6.8 fg/mL detection limits
  • Dynamic range of 6.8 - 75,000 fg/mL
  • Compatible with serum, EDTA plasma, citrate plasma, heparin plasma, and cell culture supernatants
  • Robust performance with excellent precision
  • Research use only
  • Kit size supports 5 plates
  • Multi-component storage conditions
Catalog No. NC3355616 Supplier Meso Scale Discovery Supplier No. K151E3S2
The MESO SCALE DISCOVERY S-PLEX Human TNF-α Kit is designed for the quantitative detection of human Tumor Necrosis Factor-alpha (TNF-α) with ultra sensitivity. Utilizing advanced electrochemiluminescence technology, this kit drastically improves sensitivity and dynamic range, aiming to facilitate biomarker research. Its straightforward protocol and compatibility with various sample types make it an essential tool in research labs.
  • Assay platform utilizes advanced technology
  • High sensitivity, achieving 6.8 fg/mL detection limits
  • Dynamic range of 6.8 - 75,000 fg/mL
  • Compatible with serum, EDTA plasma, citrate plasma, heparin plasma, and cell culture supernatants
  • Robust performance with excellent precision
  • Research use only
  • Kit size supports 5 plates
  • Multi-component storage conditions