Learn More
Meso Scale Discovery S-PLEX Human TNF-B Kit
SDP
Catalog No. 5000079508
Encompass
The Meso Scale Discovery K151ADPS-1 is an electrochemiluminescent immunoassay kit for the quantitative detection of phosphorylated alpha-synuclein (pSyn) in human cerebrospinal fluid and plasma The kit contains a 96-well plate coated with a monoclonal antibody to pSyn detection antibody read buffer calibrator and sample diluent The assay has a typical sensitivity of 0 04 pg/mL and a detection range of 0 05-1000 pg/mL pSyn It utilizes MSD s proprietary SULFO-TAG labels and instrumentation for ultra-sensitive detection The kit allows high-throughput quantification of pSyn a potential biomarker for Parkinson s disease and other synucleinopathies
Catalog No. 50-000-79508 Supplier Meso Scale Discovery Supplier No. K151ADPS1
The Meso Scale Discovery K151ADPS-1 is an electrochemiluminescent immunoassay kit for the quantitative detection of phosphorylated alpha-synuclein (pSyn) in human cerebrospinal fluid and plasma The kit contains a 96-well plate coated with a monoclonal antibody to pSyn detection antibody read buffer calibrator and sample diluent The assay has a typical sensitivity of 0 04 pg/mL and a detection range of 0 05-1000 pg/mL pSyn It utilizes MSD s proprietary SULFO-TAG labels and instrumentation for ultra-sensitive detection The kit allows high-throughput quantification of pSyn a potential biomarker for Parkinson s disease and other synucleinopathies