Learn More
Meso Scale Discovery S-PLEX Human TNF-B KitQP
SDP
Catalog No. 5000079497
Encompass
The Meso Scale Discovery S-PLEX Human Cytokine 30-plex Assay Kit is designed to simultaneously measure 30 human cytokines and chemokines in serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The 30 analytes measured are EGF Eotaxin-1 GM-CSF IFN- IL-10 IL-12p70 IL-13 IL-15 IL-16 IL-17A IL-17E/IL-25 IL-1 IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IP-10 MCP-1 MIP-1 MIP-1 RANTES TNF- TNF- VEGF-A VEGF-C VEGF-D The kit includes assay plates detection antibodies read buffer wash buffer diluent and calibrators
Catalog No. 50-000-79497 Supplier Meso Scale Discovery Supplier No. K151ADPS21
Add to Cart
Add to Cart
The Meso Scale Discovery S-PLEX Human Cytokine 30-plex Assay Kit is designed to simultaneously measure 30 human cytokines and chemokines in serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and low sample consumption The 30 analytes measured are EGF Eotaxin-1 GM-CSF IFN- IL-10 IL-12p70 IL-13 IL-15 IL-16 IL-17A IL-17E/IL-25 IL-1 IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IP-10 MCP-1 MIP-1 MIP-1 RANTES TNF- TNF- VEGF-A VEGF-C VEGF-D The kit includes assay plates detection antibodies read buffer wash buffer diluent and calibrators