Learn More
Meso Scale Discovery S-PLEX Human TNF-B KitQP
SDP
Catalog No. 5000079509
Encompass
The Meso Scale Discovery S-PLEX Human KinekineTM Panel 1 (22-plex) is a multiplex immunoassay that simultaneously measures 22 human cytokines chemokines growth factors and immune modulators in serum plasma and culture supernatants This assay uses electrochemiluminescence detection technology on Meso Scale Discovery s proprietary multi-array plate platform The panel includes IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IL-10 IL-12p70 IL-13 IL-15 IL-16 IL-17A IFN- TNF- TNF- GM-CSF VEGF-A CCL2 CCL3 CCL4 and CCL11 This assay allows high-throughput sensitive and reproducible quantification of multiple analytes from a small sample volume
Catalog No. 50-000-79509 Supplier Meso Scale Discovery Supplier No. K151ADPS22
The Meso Scale Discovery S-PLEX Human KinekineTM Panel 1 (22-plex) is a multiplex immunoassay that simultaneously measures 22 human cytokines chemokines growth factors and immune modulators in serum plasma and culture supernatants This assay uses electrochemiluminescence detection technology on Meso Scale Discovery s proprietary multi-array plate platform The panel includes IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IL-10 IL-12p70 IL-13 IL-15 IL-16 IL-17A IFN- TNF- TNF- GM-CSF VEGF-A CCL2 CCL3 CCL4 and CCL11 This assay allows high-throughput sensitive and reproducible quantification of multiple analytes from a small sample volume