Learn More
Meso Scale Discovery S-PLEX Human TNF- KitQP
SDP
Catalog No. 5000082434
Encompass
The S-PLEX Human TNF-alpha Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of TNF-alpha in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity and low cross-reactivity The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and controls The assay can quantify TNF-alpha concentrations between 0 3-2500 pg/mL
Catalog No. 50-000-82434 Supplier Meso Scale Discovery Supplier No. K151E3S21
The S-PLEX Human TNF-alpha Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of TNF-alpha in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity and low cross-reactivity The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and controls The assay can quantify TNF-alpha concentrations between 0 3-2500 pg/mL