Learn More
Meso Scale Discovery S-PLEX Human TSLP Kit 25 PL
SDP
Catalog No. 5000082463
Encompass
The S-PLEX Human TSLP Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human thymic stromal lymphopoietin (TSLP) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Streptavidin-HRP The assay has a sensitivity of 0 11 pg/mL and an assay range from 0 49 pg/mL to 1000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of human TSLP
Catalog No. 50-000-82463 Supplier Meso Scale Discovery Supplier No. K151D3S4
Add to Cart
Add to Cart
Request a Quote
The S-PLEX Human TSLP Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of human thymic stromal lymphopoietin (TSLP) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Streptavidin-HRP The assay has a sensitivity of 0 11 pg/mL and an assay range from 0 49 pg/mL to 1000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of human TSLP