Learn More
Meso Scale Discovery S-PLEX Human TSLP Kit, 96-Well format, LLOD 9.1 fg/mL, Dynamic Range 9.1 - 70,000 fg/mL, Human Analyte, Mouse monoclonal antibodies, For research use only
SDP
Catalog No. NC3740780
Encompass
The MESO SCALE DISCOVERY S-PLEX Human TSLP Kit is an ultrasensitive immunoassay designed for the quantitative measurement of Thymic Stromal Lymphopoietin (TSLP) in biological samples. Utilizing advanced S-PLEX technology, the kit provides exceptional sensitivity and a wide dynamic range, making it ideal for a variety of research applications in immunology and inflammation studies.
  • Immunoassay for quantifying TSLP
  • Enhanced sensitivity with a 10- to 1000-fold reduction in LLOD compared to traditional methods
  • Wide dynamic range: 9.1 - 70,000 fg/mL
  • Compatible with various MSD SECTOR and QuickPlex instruments
  • Optimized for challenging sample matrices including serum and plasma
  • Mouse monoclonal capture and detection antibodies
  • For research use only; not for diagnostic procedures
  • Multi-component storage conditions
  • Kit format: 96-Well SECTOR Plate
Catalog No. NC3740780 Supplier Meso Scale Discovery Supplier No. K151D3S2
The MESO SCALE DISCOVERY S-PLEX Human TSLP Kit is an ultrasensitive immunoassay designed for the quantitative measurement of Thymic Stromal Lymphopoietin (TSLP) in biological samples. Utilizing advanced S-PLEX technology, the kit provides exceptional sensitivity and a wide dynamic range, making it ideal for a variety of research applications in immunology and inflammation studies.
  • Immunoassay for quantifying TSLP
  • Enhanced sensitivity with a 10- to 1000-fold reduction in LLOD compared to traditional methods
  • Wide dynamic range: 9.1 - 70,000 fg/mL
  • Compatible with various MSD SECTOR and QuickPlex instruments
  • Optimized for challenging sample matrices including serum and plasma
  • Mouse monoclonal capture and detection antibodies
  • For research use only; not for diagnostic procedures
  • Multi-component storage conditions
  • Kit format: 96-Well SECTOR Plate