Learn More
Meso Scale Discovery S-PLEX Neurology Panel 1 NHP, multiplex immunoassay, measures GFAP, Neurofilament L, total Tau, for NHP serum/plasma/CSF samples, 96-well plate
SDP
Catalog No. NC2949130
Encompass
The MESO SCALE DISCOVERY S-PLEX Neurology Panel 1 NHP is an ultrasensitive multiplex immunoassay that facilitates the quantitative measurement of critical neurological biomarkers such as GFAP, Neurofilament L, and total Tau in non-human primate serum, plasma, and cerebrospinal fluid (CSF) samples. This innovative kit enhances sensitivity and ensures robust performance for neuroinflammation and neurodegeneration research.
  • Offers ultrasensitive detection in the low femtogram/mL range.
  • Allows multiplexed analysis of multiple biomarkers from a single sample.
  • Compatible with NHP serum, plasma, and CSF.
  • Features rigorous quality control and excellent matrix tolerance.
  • Optimized for studies on brain injury and various neurological disorders.
Catalog No. NC2949130 Supplier Meso Scale Discovery Supplier No. K15640S1
The MESO SCALE DISCOVERY S-PLEX Neurology Panel 1 NHP is an ultrasensitive multiplex immunoassay that facilitates the quantitative measurement of critical neurological biomarkers such as GFAP, Neurofilament L, and total Tau in non-human primate serum, plasma, and cerebrospinal fluid (CSF) samples. This innovative kit enhances sensitivity and ensures robust performance for neuroinflammation and neurodegeneration research.
  • Offers ultrasensitive detection in the low femtogram/mL range.
  • Allows multiplexed analysis of multiple biomarkers from a single sample.
  • Compatible with NHP serum, plasma, and CSF.
  • Features rigorous quality control and excellent matrix tolerance.
  • Optimized for studies on brain injury and various neurological disorders.