Learn More
Meso Scale Discovery S-PLEX NHP CTLA-4 Kit
SDP
Catalog No. 5000081029
Encompass
Meso Scale Diagnostics offers a wide range of multiplex immunoassay kits that allow researchers to measure biomarkers cytokines chemokines growth factors immunoglobulins and more from biological samples such as serum plasma cell culture supernatants and other sample types Their S-PLEX assays use electrochemiluminescence detection for high sensitivity broad dynamic range and minimal matrix effects With the ability to multiplex up to 10 analytes per well in a standard 96-well plate format their assays provide an efficient and cost-effective solution for biomarker analysis The assays are available in both off-the-shelf and custom panel formats to meet diverse research needs
Catalog No. 50-000-81029 Supplier Meso Scale Discovery Supplier No. K156AGLS1
Meso Scale Diagnostics offers a wide range of multiplex immunoassay kits that allow researchers to measure biomarkers cytokines chemokines growth factors immunoglobulins and more from biological samples such as serum plasma cell culture supernatants and other sample types Their S-PLEX assays use electrochemiluminescence detection for high sensitivity broad dynamic range and minimal matrix effects With the ability to multiplex up to 10 analytes per well in a standard 96-well plate format their assays provide an efficient and cost-effective solution for biomarker analysis The assays are available in both off-the-shelf and custom panel formats to meet diverse research needs