Learn More
Meso Scale Discovery S-PLEX NHP CTLA-4 KitQP
SDP
Catalog No. 5000081018
Encompass
The Meso Scale Discovery CTLA-4 Assay Kit is a sensitive electrochemiluminescent immunoassay for quantifying soluble CTLA-4 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for superior sensitivity and minimal matrix interference The detection range is 0 156 to 50 ng/mL The kit includes a 96-well plate detection antibody calibrators and diluent Total assay time is 3 5 hours enabling sensitive and reproducible CTLA-4 measurement for research
Catalog No. 50-000-81018 Supplier Meso Scale Discovery Supplier No. K156AGLS22
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery CTLA-4 Assay Kit is a sensitive electrochemiluminescent immunoassay for quantifying soluble CTLA-4 in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for superior sensitivity and minimal matrix interference The detection range is 0 156 to 50 ng/mL The kit includes a 96-well plate detection antibody calibrators and diluent Total assay time is 3 5 hours enabling sensitive and reproducible CTLA-4 measurement for research