Learn More
Meso Scale Discovery S-PLEX NHP G-CSF Kit 25 PL
SDP
Catalog No. 5000081275
Encompass
The Meso Scale Discovery S-PLEX Human IL-6 Kit is a multiplex immunoassay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and controls It requires a Meso Scale Discovery instrument takes about 3 5 hours and detects IL-6 from 0 06 to 1500 pg/mL ensuring reliable quantification across a wide range
Catalog No. 50-000-81275 Supplier Meso Scale Discovery Supplier No. K156K3S4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery S-PLEX Human IL-6 Kit is a multiplex immunoassay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and controls It requires a Meso Scale Discovery instrument takes about 3 5 hours and detects IL-6 from 0 06 to 1500 pg/mL ensuring reliable quantification across a wide range