Learn More
Meso Scale Discovery S-PLEX NHP G-CSF Kit
SDP
Catalog No. 5000081091
Encompass
The Meso Scale Discovery G-CSF Kit is an electrochemiluminescent immunoassay for measuring granulocyte colony stimulating factor (G-CSF) in human serum plasma and tissue culture media It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents It measures G-CSF from 78 to 10 000 pg/mL with an LLOQ of 64 pg/mL and takes about 3 5 hours to complete
Catalog No. 50-000-81091 Supplier Meso Scale Discovery Supplier No. K156K3S2
The Meso Scale Discovery G-CSF Kit is an electrochemiluminescent immunoassay for measuring granulocyte colony stimulating factor (G-CSF) in human serum plasma and tissue culture media It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents It measures G-CSF from 78 to 10 000 pg/mL with an LLOQ of 64 pg/mL and takes about 3 5 hours to complete