Learn More
Meso Scale Discovery S-PLEX NHP G-CSF KitQP
SDP
Catalog No. 5000079970
Encompass
The S-Plex NHP-G CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of neurofilament light chain (NfL) glial fibrillary acidic protein (GFAP) and neurogranin (Ng) in human cerebrospinal fluid (CSF) The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls This assay allows for the simultaneous measurement of NfL GFAP and Ng biomarkers which may aid in the assessment of neurodegeneration The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range
Catalog No. 50-000-79970 Supplier Meso Scale Discovery Supplier No. K156K3S22
The S-Plex NHP-G CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of neurofilament light chain (NfL) glial fibrillary acidic protein (GFAP) and neurogranin (Ng) in human cerebrospinal fluid (CSF) The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls This assay allows for the simultaneous measurement of NfL GFAP and Ng biomarkers which may aid in the assessment of neurodegeneration The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range