Learn More
Meso Scale Discovery S-PLEX NHP G-CSF KitQP
SDP
Catalog No. 5000081058
Encompass
The Meso Scale Discovery G-CSF Kit is an electrochemiluminescent immunoassay for the quantitative measurement of granulocyte colony stimulating factor (G-CSF) in human serum plasma and tissue culture media The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the assay The kit can quantitatively measure G-CSF over a range of 156 - 20 000 pg/mL with an LLOQ of 64 5 pg/mL
Catalog No. 50-000-81058 Supplier Meso Scale Discovery Supplier No. K156K3S21
The Meso Scale Discovery G-CSF Kit is an electrochemiluminescent immunoassay for the quantitative measurement of granulocyte colony stimulating factor (G-CSF) in human serum plasma and tissue culture media The assay uses MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide high sensitivity wide dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to complete the assay The kit can quantitatively measure G-CSF over a range of 156 - 20 000 pg/mL with an LLOQ of 64 5 pg/mL