Learn More
Meso Scale Discovery S-PLEX NHP IL-1B Kit
SDP
Catalog No. 5000081017
Encompass
The Meso Scale Discovery S-PLEX NHP IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1 in non-human primate serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager 2400 or SECTOR S 600 plate reader and provides a detection range of 0 05-1000 pg/mL with an assay time of 3 5 hours This multiplex assay allows for the simultaneous measurement of IL-1 with other biomarkers The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators
Catalog No. 50-000-81017 Supplier Meso Scale Discovery Supplier No. K156ADSS1
The Meso Scale Discovery S-PLEX NHP IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-1 in non-human primate serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager 2400 or SECTOR S 600 plate reader and provides a detection range of 0 05-1000 pg/mL with an assay time of 3 5 hours This multiplex assay allows for the simultaneous measurement of IL-1 with other biomarkers The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent and calibrators