Learn More
Meso Scale Discovery S-PLEX NHP IL-1B Kit
SDP
Catalog No. 5000081089
Encompass
Meso Scale Discovery offers multiplex immunoassay kits for measuring multiple protein biomarkers in samples like serum plasma tissue homogenates and cell lysates Their S-PLEX assays provide high sensitivity broad dynamic range and low sample volume requirements With minimal dilution they measure biomarkers from pg/mL to g/mL They offer panels for research in immunology oncology neurobiology and cardiovascular disease The electrochemiluminescent detection ensures high sensitivity low background and wide dynamic range
Catalog No. 50-000-81089 Supplier Meso Scale Discovery Supplier No. K156ADSS2
Meso Scale Discovery offers multiplex immunoassay kits for measuring multiple protein biomarkers in samples like serum plasma tissue homogenates and cell lysates Their S-PLEX assays provide high sensitivity broad dynamic range and low sample volume requirements With minimal dilution they measure biomarkers from pg/mL to g/mL They offer panels for research in immunology oncology neurobiology and cardiovascular disease The electrochemiluminescent detection ensures high sensitivity low background and wide dynamic range