Learn More
Meso Scale Discovery S-PLEX NHP IL-1B KitQP 25 PL
SDP
Catalog No. 5000080948
Encompass
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a highly sensitive and quantitative assay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay utilizes MSD s proprietary V-PLEX technology and provides accurate and reproducible results in just 3 5 hours The kit includes a 96-well plate coated with capture antibodies against IL-6 detection antibodies labeled with an electrochemiluminescent compound calibrators controls assay and wash buffers and plate seals The kit can quantify IL-6 over a range of 0 2-2500 pg/mL and requires just 25 L of sample per well
Catalog No. 50-000-80948 Supplier Meso Scale Discovery Supplier No. K156ADSS24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-6 Kit is a highly sensitive and quantitative assay for measuring human interleukin 6 (IL-6) in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay utilizes MSD s proprietary V-PLEX technology and provides accurate and reproducible results in just 3 5 hours The kit includes a 96-well plate coated with capture antibodies against IL-6 detection antibodies labeled with an electrochemiluminescent compound calibrators controls assay and wash buffers and plate seals The kit can quantify IL-6 over a range of 0 2-2500 pg/mL and requires just 25 L of sample per well