Learn More
Meso Scale Discovery S-PLEX NHP IL-1B KitQP
SDP
Catalog No. 5000081019
Encompass
The S-PLEX NHP IL-1beta Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of IL-1beta in non-human primate serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label assay diluent wash buffer read buffer and calibrator The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-81019 Supplier Meso Scale Discovery Supplier No. K156ADSS22
The S-PLEX NHP IL-1beta Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of IL-1beta in non-human primate serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label assay diluent wash buffer read buffer and calibrator The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL