Learn More
Meso Scale Discovery S-PLEX NHP IL-2 KitQP
SDP
Catalog No. 5000078616
Encompass
The Meso Scale Discovery S-PLEX Human IL-6 Kit is a multiplex immunoassay for measuring human interleukin-6 (IL-6) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and controls The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument It detects IL-6 from 0 06 to 10 000 pg/mL ensuring reliable quantification across a wide concentration range
Catalog No. 50-000-78616 Supplier Meso Scale Discovery Supplier No. K156Z2S22
The Meso Scale Discovery S-PLEX Human IL-6 Kit is a multiplex immunoassay for measuring human interleukin-6 (IL-6) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and controls The assay takes about 3 5 hours and requires a Meso Scale Discovery instrument It detects IL-6 from 0 06 to 10 000 pg/mL ensuring reliable quantification across a wide concentration range