Learn More
Meso Scale Discovery S-PLEX NHP IL-2 KitQP
SDP
Catalog No. 5000080777
Encompass
Meso Scale Discovery offers a wide range of multiplex immunoassay kits that enable simultaneous quantitative measurement of multiple protein biomarkers in diverse sample types such as serum plasma tissue lysates and cell culture supernatants Their S-PLEX assays utilize electrochemiluminescence for high sensitivity and broad dynamic range With minimal sample volume requirements rapid assay times and minimal hands-on time their kits are well-suited for biomarker research drug development and clinical research applications Their catalog contains S-PLEX assays for cytokines chemokines growth factors immunoglobulins metabolic markers and more
Catalog No. 50-000-80777 Supplier Meso Scale Discovery Supplier No. K156Z2S21
Meso Scale Discovery offers a wide range of multiplex immunoassay kits that enable simultaneous quantitative measurement of multiple protein biomarkers in diverse sample types such as serum plasma tissue lysates and cell culture supernatants Their S-PLEX assays utilize electrochemiluminescence for high sensitivity and broad dynamic range With minimal sample volume requirements rapid assay times and minimal hands-on time their kits are well-suited for biomarker research drug development and clinical research applications Their catalog contains S-PLEX assays for cytokines chemokines growth factors immunoglobulins metabolic markers and more