Learn More
Meso Scale Discovery S-PLEX NHP IL-5 Kit
SDP
Catalog No. 5000080836
Encompass
Meso Scale Diagnostics offers a wide range of multiplex immunoassay kits that allow researchers to simultaneously measure multiple protein biomarkers from a single sample The S-PLEX assays utilize electrochemiluminescent detection for high sensitivity and broad dynamic range Kits are available to measure cytokines chemokines growth factors immunoglobulins and other proteins across sample types including serum plasma cell culture supernatants and more With minimal sample volume requirements and high-throughput compatible plates these assays provide an efficient way to generate reproducible quantitative data for biomarker studies
Catalog No. 50-000-80836 Supplier Meso Scale Discovery Supplier No. K156J3S2
Add to Cart
Add to Cart
Meso Scale Diagnostics offers a wide range of multiplex immunoassay kits that allow researchers to simultaneously measure multiple protein biomarkers from a single sample The S-PLEX assays utilize electrochemiluminescent detection for high sensitivity and broad dynamic range Kits are available to measure cytokines chemokines growth factors immunoglobulins and other proteins across sample types including serum plasma cell culture supernatants and more With minimal sample volume requirements and high-throughput compatible plates these assays provide an efficient way to generate reproducible quantitative data for biomarker studies