Learn More
Meso Scale Discovery S-PLEX NHP IL-5 KitQP
SDP
Catalog No. 5000080946
Encompass
The Mesoscale K156J3S-22 is a 22-pin DIP socket designed for use with microcontrollers It is constructed with phosphor bronze contacts and a glass filled polyester insulator for durability The socket accepts standard 0 3 (7 62mm) lead spacing and has a current rating of 1A per pin With a profile height of 0 335 (8 5mm) this socket allows easy insertion and removal of ICs while providing reliable electrical connection The K156J3S-22 socket is suitable for prototyping testing and low-to-medium volume production applications
Catalog No. 50-000-80946 Supplier Meso Scale Discovery Supplier No. K156J3S22
The Mesoscale K156J3S-22 is a 22-pin DIP socket designed for use with microcontrollers It is constructed with phosphor bronze contacts and a glass filled polyester insulator for durability The socket accepts standard 0 3 (7 62mm) lead spacing and has a current rating of 1A per pin With a profile height of 0 335 (8 5mm) this socket allows easy insertion and removal of ICs while providing reliable electrical connection The K156J3S-22 socket is suitable for prototyping testing and low-to-medium volume production applications