Learn More
Meso Scale Discovery S-PLEX NHP IL-6 KitQP 25 PL
SDP
Catalog No. 5000078613
Encompass
The S-PLEX NHP IL-6 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants The assay has high sensitivity wide dynamic range and requires low sample volume The kit includes a 96-well S-PLEX plate coated with IL-6 capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78613 Supplier Meso Scale Discovery Supplier No. K156B3S24
Add to Cart
Add to Cart
Request a Quote
The S-PLEX NHP IL-6 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants The assay has high sensitivity wide dynamic range and requires low sample volume The kit includes a 96-well S-PLEX plate coated with IL-6 capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator The assay takes approximately 3 5 hours to complete