Learn More
Meso Scale Discovery S-PLEX NHP IL-6 KitQP
SDP
Catalog No. 5000081033
Encompass
Meso Scale Discovery offers a range of multiplex immunoassay kits that enable simultaneous quantitative measurement of multiple protein biomarkers in diverse sample types such as serum plasma tissue lysates and cell culture supernatants Their S-PLEX assays use electrochemiluminescence for high sensitivity and broad dynamic range With minimal sample volume requirements rapid assay times and standard curves for each biomarker S-PLEX kits allow efficient and accurate quantification of up to 10 proteins per well The assays are available in a variety of pre-designed and custom panels optimized for cancer immunology neurology and cardiovascular research
Catalog No. 50-000-81033 Supplier Meso Scale Discovery Supplier No. K156B3S21
Meso Scale Discovery offers a range of multiplex immunoassay kits that enable simultaneous quantitative measurement of multiple protein biomarkers in diverse sample types such as serum plasma tissue lysates and cell culture supernatants Their S-PLEX assays use electrochemiluminescence for high sensitivity and broad dynamic range With minimal sample volume requirements rapid assay times and standard curves for each biomarker S-PLEX kits allow efficient and accurate quantification of up to 10 proteins per well The assays are available in a variety of pre-designed and custom panels optimized for cancer immunology neurology and cardiovascular research