Learn More
Meso Scale Discovery S-PLEX NHP IL-6 KitQP
SDP
Catalog No. 5000081057
Encompass
The Meso Scale Discovery S-PLEX NHP IL-6 Kit is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants The assay has high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent and wash buffer The assay takes approximately 3 5 hours to complete with only 2 hours hands-on time The kit has a lower limit of detection of 0 10 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81057 Supplier Meso Scale Discovery Supplier No. K156B3S22
The Meso Scale Discovery S-PLEX NHP IL-6 Kit is an immunoassay that uses electrochemiluminescence (ECL) technology for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants The assay has high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators diluent and wash buffer The assay takes approximately 3 5 hours to complete with only 2 hours hands-on time The kit has a lower limit of detection of 0 10 pg/mL and an upper limit of quantification of 10 000 pg/mL