Learn More
Meso Scale Discovery S-PLEX NHP Neurofilament L Kit, 96-well plate format, Dynamic Range: 1,700 - 1,400,000 fg/mL, Sensitivity: 1,700 fg/mL, For research use only
SDP
Catalog No. NC3353151
Encompass
S-PLEX NHP Neurofilament L Kit, 96-well plate format, Dynamic Range: 1,700 - 1,400,000 fg/mL, Sensitivity: 1,700 fg/mL, For research use only
Catalog No. NC3353151 Supplier Meso Scale Discovery Supplier No. K156AKGS1
S-PLEX NHP Neurofilament L Kit, 96-well plate format, Dynamic Range: 1,700 - 1,400,000 fg/mL, Sensitivity: 1,700 fg/mL, For research use only