Learn More
Meso Scale Discovery S-PLEX NHP Tau pT181 Kit
SDP
Catalog No. 5000081061
Encompass
The K156AGMS-1 is a high-performance thermoelectric module from Mesoscale that uses bismuth telluride alloys to provide efficient heating and cooling It measures 40 x 40 x 3 4mm and can achieve a maximum temperature differential of 70 C With a thermal conductivity of 1 5 W/m-K it provides 156 Watts of cooling capacity at T 0 C when powered by a 6 4V DC supply The module has a no-load voltage of 3 4V and internal resistance of 1 8 ohms It can operate from -40 C to 200 C ambient temperature The K156AGMS-1 module is designed for precise temperature control applications
Catalog No. 50-000-81061 Supplier Meso Scale Discovery Supplier No. K156AGMS1
The K156AGMS-1 is a high-performance thermoelectric module from Mesoscale that uses bismuth telluride alloys to provide efficient heating and cooling It measures 40 x 40 x 3 4mm and can achieve a maximum temperature differential of 70 C With a thermal conductivity of 1 5 W/m-K it provides 156 Watts of cooling capacity at T 0 C when powered by a 6 4V DC supply The module has a no-load voltage of 3 4V and internal resistance of 1 8 ohms It can operate from -40 C to 200 C ambient temperature The K156AGMS-1 module is designed for precise temperature control applications