Learn More
Meso Scale Discovery S-PLEX NHP Tau pT231 Kit
SDP
Catalog No. 5000080855
Encompass
The Meso Scale Discovery Tau (pT231) Kit is an electrochemiluminescent immunoassay for measuring phosphorylated Tau protein at Threonine 231 in human samples like serum plasma and CSF It uses SULFO-TAG labels that emit light when stimulated The kit includes a 96-well plate with a capture antibody for pTau231 detection antibody calibrator and diluent It has a sensitivity of 1-2 pg/mL and a range up to 10 000 pg/mL allowing high-throughput analysis of Alzheimer s-related biomarkers
Catalog No. 50-000-80855 Supplier Meso Scale Discovery Supplier No. K156AGNS1
The Meso Scale Discovery Tau (pT231) Kit is an electrochemiluminescent immunoassay for measuring phosphorylated Tau protein at Threonine 231 in human samples like serum plasma and CSF It uses SULFO-TAG labels that emit light when stimulated The kit includes a 96-well plate with a capture antibody for pTau231 detection antibody calibrator and diluent It has a sensitivity of 1-2 pg/mL and a range up to 10 000 pg/mL allowing high-throughput analysis of Alzheimer s-related biomarkers