Learn More
Meso Scale Discovery S-PLEX NHP TNF-B Kit 25 PL
SDP
Catalog No. 5000079304
Encompass
Meso Scale Discovery offers multiplex immunoassay kits for simultaneous measurement of multiple protein biomarkers in samples like serum plasma and tissue homogenates Their S-PLEX assays use electrochemiluminescent detection for high sensitivity and broad dynamic range With minimal sample volume rapid assay times and up to 10-plex multiplexing MSD s assays are ideal for biomarker research drug development and diagnostics Their robust performance and streamlined workflow enable researchers to obtain more data from limited samples
Catalog No. 50-000-79304 Supplier Meso Scale Discovery Supplier No. K156ADPS4
Add to Cart
Add to Cart
Request a Quote
Meso Scale Discovery offers multiplex immunoassay kits for simultaneous measurement of multiple protein biomarkers in samples like serum plasma and tissue homogenates Their S-PLEX assays use electrochemiluminescent detection for high sensitivity and broad dynamic range With minimal sample volume rapid assay times and up to 10-plex multiplexing MSD s assays are ideal for biomarker research drug development and diagnostics Their robust performance and streamlined workflow enable researchers to obtain more data from limited samples