Learn More
Meso Scale Discovery S-PLEX NHP TNF-B Kit
SDP
Catalog No. 5000078612
Encompass
The Meso Scale Discovery S-PLEX Human Kits are multiplex immunoassays that enable simultaneous quantitative measurement of multiple protein biomarkers in human biological samples such as serum plasma and cell culture supernatants The K156ADPS-2 kit allows for the simultaneous measurement of 156 biomarkers including cytokines chemokines growth factors immunoglobulins acute phase proteins and hormones It utilizes MSD s proprietary electrochemiluminescent detection technology for high sensitivity wide dynamic range and low sample volume requirements This multiplex assay kit provides a high-throughput method for biomarker profiling and research
Catalog No. 50-000-78612 Supplier Meso Scale Discovery Supplier No. K156ADPS2
Add to Cart
Add to Cart
The Meso Scale Discovery S-PLEX Human Kits are multiplex immunoassays that enable simultaneous quantitative measurement of multiple protein biomarkers in human biological samples such as serum plasma and cell culture supernatants The K156ADPS-2 kit allows for the simultaneous measurement of 156 biomarkers including cytokines chemokines growth factors immunoglobulins acute phase proteins and hormones It utilizes MSD s proprietary electrochemiluminescent detection technology for high sensitivity wide dynamic range and low sample volume requirements This multiplex assay kit provides a high-throughput method for biomarker profiling and research