Learn More
Meso Scale Discovery S-PLEX SARS-CoV-2 N Kit
SDP
Catalog No. 5000078815
Encompass
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel for the simultaneous qualitative detection of nucleocapsid protein antigens to SARS-CoV-2 in serum plasma and saliva This assay utilizes electrochemiluminescence (ECL) on the SECTOR Imager 2400 platform The kit includes a plate coated with capture antibodies detection antibodies conjugated to ruthenium and calibrators and controls needed to run the assay Results are obtained within 3 hours This is an in vitro diagnostic test for aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-78815 Supplier Meso Scale Discovery Supplier No. K150ADHS2
Add to Cart
Add to Cart
Request a Quote
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel for the simultaneous qualitative detection of nucleocapsid protein antigens to SARS-CoV-2 in serum plasma and saliva This assay utilizes electrochemiluminescence (ECL) on the SECTOR Imager 2400 platform The kit includes a plate coated with capture antibodies detection antibodies conjugated to ruthenium and calibrators and controls needed to run the assay Results are obtained within 3 hours This is an in vitro diagnostic test for aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection