Learn More
Meso Scale Discovery S-PLEX SARS-CoV-2 N KitQP
SDP
Catalog No. 5000079225
Encompass
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel for the simultaneous qualitative detection of nucleocapsid protein antigens from SARS-CoV-2 in human serum plasma and saliva This assay utilizes electrochemiluminescent detection technology on the SECTOR Imager 2400 platform The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium assay diluent wash buffer read buffer and calibrators This is an in vitro diagnostic test for authorized laboratories With high sensitivity and specificity it can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-79225 Supplier Meso Scale Discovery Supplier No. K150ADHS21
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel for the simultaneous qualitative detection of nucleocapsid protein antigens from SARS-CoV-2 in human serum plasma and saliva This assay utilizes electrochemiluminescent detection technology on the SECTOR Imager 2400 platform The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium assay diluent wash buffer read buffer and calibrators This is an in vitro diagnostic test for authorized laboratories With high sensitivity and specificity it can aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection