Learn More
Meso Scale Discovery S-PLEX SARS-CoV-2 N KitQP
SDP
Catalog No. 5000079226
Encompass
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel that detects antibodies to the nucleocapsid protein of SARS-CoV-2 in serum and plasma samples This qualitative assay uses electrochemiluminescence detection technology and provides results in under 3 hours The kit includes all necessary reagents and can process up to 30 samples per run This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-79226 Supplier Meso Scale Discovery Supplier No. K150ADHS22
Add to Cart
Add to Cart
Request a Quote
The S-PLEX SARS-CoV-2 N Kit is a multiplex immunoassay panel that detects antibodies to the nucleocapsid protein of SARS-CoV-2 in serum and plasma samples This qualitative assay uses electrochemiluminescence detection technology and provides results in under 3 hours The kit includes all necessary reagents and can process up to 30 samples per run This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection