Learn More
Meso Scale Discovery S-PLEX SARS-CoV-2 Spike Kit
SDP
Catalog No. 5000080527
Encompass
The S-PLEX SARS-CoV-2 Spike Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that quantitatively measures antibodies specific to the SARS-CoV-2 Spike protein This multiplex assay allows for the simultaneous detection of IgG IgM and IgA antibodies in serum plasma or whole blood samples With high sensitivity and specificity it can aid in understanding immune responses and seroprevalence studies related to COVID-19 infection and vaccination The kit includes a 96-well plate calibrators controls diluent and reagents for up to 88 tests
Catalog No. 50-000-80527 Supplier Meso Scale Discovery Supplier No. K150ADJS1
The S-PLEX SARS-CoV-2 Spike Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that quantitatively measures antibodies specific to the SARS-CoV-2 Spike protein This multiplex assay allows for the simultaneous detection of IgG IgM and IgA antibodies in serum plasma or whole blood samples With high sensitivity and specificity it can aid in understanding immune responses and seroprevalence studies related to COVID-19 infection and vaccination The kit includes a 96-well plate calibrators controls diluent and reagents for up to 88 tests