Learn More
Meso Scale Discovery S-PLEX SARS-CoV-2 Spike KitQP
SDP
Catalog No. 5000078283
Encompass
The S-PLEX SARS-CoV-2 Spike Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that detects antibodies to the SARS-CoV-2 spike protein in human serum and plasma This multiplex kit includes a 96-well plate coated with the SARS-CoV-2 spike protein as well as calibrators and controls The assay has high sensitivity and specificity for detecting antibodies that bind to the receptor binding domain of the SARS-CoV-2 spike protein Results are obtained in just a few hours using an MSD reader This kit can aid in understanding immune responses to SARS-CoV-2 infection and vaccination
Catalog No. 50-000-78283 Supplier Meso Scale Discovery Supplier No. K150ADJS21
The S-PLEX SARS-CoV-2 Spike Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that detects antibodies to the SARS-CoV-2 spike protein in human serum and plasma This multiplex kit includes a 96-well plate coated with the SARS-CoV-2 spike protein as well as calibrators and controls The assay has high sensitivity and specificity for detecting antibodies that bind to the receptor binding domain of the SARS-CoV-2 spike protein Results are obtained in just a few hours using an MSD reader This kit can aid in understanding immune responses to SARS-CoV-2 infection and vaccination